Product Details
- SNP ID
-
rs2270400
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:17202626 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATAACTGACCCGAAGTAACAGAA[A/C]TTTCCCTTTCCTAACCTTCTTTAGT
- Phenotype
-
MIM: 607273
MIM: 612935
MIM: 614960
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FLCN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75836479] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FLCN
- Gene Name
- folliculin
There are no transcripts associated with this gene.
- Gene
- MPRIP
- Gene Name
- myosin phosphatase Rho interacting protein
There are no transcripts associated with this gene.
- Gene
- PLD6
- Gene Name
- phospholipase D family member 6
View Full Product Details