Product Details

SNP ID
rs2304222
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39500563 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAACTCTGGCCTTCATTGAGTACTT[A/G]CCCTAACCACTGTCTTTCATCTTTC
Phenotype
MIM: 602768 MIM: 607381
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DLL3 PubMed Links

Gene Details

Gene
DLL3
Gene Name
delta like canonical Notch ligand 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016941.3 Intron NP_058637.1
NM_203486.2 Intron NP_982353.1
Gene
TIMM50
Gene Name
translocase of inner mitochondrial membrane 50
There are no transcripts associated with this gene.

View Full Product Details