Product Details

SNP ID
rs2454041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:103021284 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTGGTCAGGCCGAAGCATGAGCT[C/G]CGGTCGCTGCCTGGGAGGGACGCGG
Phenotype
MIM: 603097
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP6V1C1 PubMed Links

Gene Details

Gene
ATP6V1C1
Gene Name
ATPase H+ transporting V1 subunit C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001695.4 Intron NP_001686.1

View Full Product Details