Product Details

SNP ID
rs2254132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:32298296 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAGATAAACAGCCTTTTGAGTCT[C/G]AACAAATGCAGCCGGCTGGTTGGTT
Phenotype
MIM: 609196
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC105372779 PubMed Links

Gene Details

Gene
LOC105372779
Gene Name
uncharacterized LOC105372779
There are no transcripts associated with this gene.

Gene
MRAP
Gene Name
melanocortin 2 receptor accessory protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285394.1 Intron NP_001272323.1
NM_178817.3 Intron NP_848932.1
NM_206898.1 Intron NP_996781.1
XM_006724028.3 Intron XP_006724091.1
XM_017028407.1 Intron XP_016883896.1

View Full Product Details