Product Details

SNP ID
rs2239971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:96123069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGTGTAAATCTGCATACTACAC[A/G]CTTCCATTTATATTACTGAAGGAGT
Phenotype
MIM: 603859
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A13 PubMed Links

Gene Details

Gene
SLC25A13
Gene Name
solute carrier family 25 member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160210.1 Intron NP_001153682.1
NM_014251.2 Intron NP_055066.1
XM_006715831.3 Intron XP_006715894.1
XM_011515727.2 Intron XP_011514029.1
XM_017011663.1 Intron XP_016867152.1
XM_017011664.1 Intron XP_016867153.1
XM_017011665.1 Intron XP_016867154.1

View Full Product Details