Product Details

SNP ID
rs2273837
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:22873373 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGGCACCTTACAGAGGCCCCTGGTC[C/T]GGGACAGCCGCACCTCCCCTGCCAA
Phenotype
MIM: 609921 MIM: 616955
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRP10 PubMed Links

Gene Details

Gene
LRP10
Gene Name
LDL receptor related protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005267510.1 833 Silent Mutation CGG,TGG R48W XP_005267567.1
Gene
REM2
Gene Name
RRAD and GEM like GTPase 2
There are no transcripts associated with this gene.

View Full Product Details