Product Details

SNP ID
rs2274738
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:85654215 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GCAAAAGCACTGCTCACAGGGACAC[A/G]CAAACCGCGTCATGTCCCAGGCAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNHIT6 PubMed Links

Gene Details

Gene
ZNHIT6
Gene Name
zinc finger HIT-type containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170670.1 Intron NP_001164141.1
NM_017953.3 Intron NP_060423.3
XM_011541614.2 Intron XP_011539916.1

View Full Product Details