Product Details

SNP ID
rs3025123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:24284326 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGTGGGAGCGAACGGGGCGCCCG[C/G]GAACTTGCTAGAGACGCAGCCTCCC
Phenotype
MIM: 162640
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC107986777 PubMed Links

Gene Details

Gene
LOC107986777
Gene Name
uncharacterized LOC107986777
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017012910.1 Intron XP_016868399.1
XM_017012911.1 Intron XP_016868400.1
Gene
NPY
Gene Name
neuropeptide Y
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000905.3 Intron NP_000896.1

View Full Product Details