Product Details

SNP ID
rs1805635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:180984199 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTATCCAAGTTGCCAGGCCAGTTG[C/T]CTGTACCTGGAACAGCCTTTCCACC
Phenotype
MIM: 608977 MIM: 600819
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC19 PubMed Links

Gene Details

Gene
DNAJC19
Gene Name
DnaJ heat shock protein family (Hsp40) member C19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190233.1 964 UTR 3 NP_001177162.1
NM_145261.3 964 UTR 3 NP_660304.1
Gene
FXR1
Gene Name
FMR1 autosomal homolog 1
There are no transcripts associated with this gene.

View Full Product Details