Product Details

SNP ID
rs2738241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:39371818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATTCAACTACCTAGAAATAGTT[A/C]CCTCCAGTTTAGCACATTTAGGTAT
Phenotype
MIM: 120940 MIM: 601236
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C9 PubMed Links

Gene Details

Gene
C9
Gene Name
complement component 9
There are no transcripts associated with this gene.

Gene
DAB2
Gene Name
DAB2, clathrin adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244871.1 4394 UTR 3 NP_001231800.1
NM_001343.3 4394 UTR 3 NP_001334.2

View Full Product Details