Product Details

SNP ID
rs2230344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98532142 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTCCTTACACCTCTGTCTTCCTT[C/T]CAGTCTTTTACACAGCTGTGTTCCT
Phenotype
MIM: 601166
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDND1 PubMed Links

Gene Details

Gene
CLDND1
Gene Name
claudin domain containing 1
There are no transcripts associated with this gene.

Gene
GPR15
Gene Name
G protein-coupled receptor 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005290.3 244 Missense Mutation CCA,TCA P37S NP_005281.1

View Full Product Details