Product Details
- SNP ID
-
rs28652843
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:5017302 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TCCCTGGGGCCCTCCCTGATGCTAC[A/G]ACATGCACAGTACTTTCTTGTGTCA
- Phenotype
-
MIM: 607930
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CYTL1
PubMed Links
Gene Details
- Gene
- CYTL1
- Gene Name
- cytokine like 1
View Full Product Details