Product Details
- SNP ID
-
rs2241883
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:88124547 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTCACAGACTTGATGTTTTTGAAAG[T/C]TGTCACCAGTTTATTGTCACCTTCC
- Phenotype
-
MIM: 134650
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FABP1
PubMed Links
Gene Details
- Gene
- FABP1
- Gene Name
- fatty acid binding protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001443.2 |
394 |
Missense Mutation |
ACT,GCT |
T94A |
NP_001434.1 |
View Full Product Details