Product Details
- SNP ID
-
rs114286667
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:7535654 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTACAACTCCCGGCGTGCTCAGCG[C/G]GCATTACGTGGTCTGGCGATAACGC
- Phenotype
-
MIM: 605248
MIM: 603197
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MCOLN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74956393] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MCOLN1
- Gene Name
- mucolipin 1
There are no transcripts associated with this gene.
- Gene
- PNPLA6
- Gene Name
- patatin like phospholipase domain containing 6
View Full Product Details