Product Details

SNP ID
rs142534162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:127589258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACTTTTCTGGAGCTGACCCTGA[C/T]ACCAGATACAGCTATTTGAGAAGTG
Phenotype
MIM: 103188 MIM: 615800 MIM: 607418
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARF5 PubMed Links

Gene Details

Gene
ARF5
Gene Name
ADP ribosylation factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001662.3 Intron NP_001653.1
Gene
FSCN3
Gene Name
fascin actin-bundling protein 3
There are no transcripts associated with this gene.

Gene
GCC1
Gene Name
GRIP and coiled-coil domain containing 1
There are no transcripts associated with this gene.

View Full Product Details