Product Details

SNP ID
rs35529250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:40426074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCAAATGGCACGTAACTGGCCC[T/C]GTAGATCCCGGCAGTAGGAATTCTC
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
RBM47 PubMed Links

Gene Details

Gene
RBM47
Gene Name
RNA binding motif protein 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098634.1 2443 Missense Mutation NP_001092104.1
NM_019027.3 2443 Missense Mutation NP_061900.2
XM_005248103.3 2443 Missense Mutation XP_005248160.1
XM_005248107.3 2443 Missense Mutation XP_005248164.1
XM_005248108.3 2443 Missense Mutation XP_005248165.1
XM_005248109.4 2443 Missense Mutation XP_005248166.1
XM_011513704.2 2443 Missense Mutation XP_011512006.1
XM_011513707.2 2443 Missense Mutation XP_011512009.1
XM_011513708.2 2443 Missense Mutation XP_011512010.1
XM_017008304.1 2443 Missense Mutation XP_016863793.1
XM_017008305.1 2443 Missense Mutation XP_016863794.1
XM_017008306.1 2443 Missense Mutation XP_016863795.1
XM_017008307.1 2443 Missense Mutation XP_016863796.1
XM_017008308.1 2443 Missense Mutation XP_016863797.1
XM_017008309.1 2443 Missense Mutation XP_016863798.1
XM_017008310.1 2443 Missense Mutation XP_016863799.1

View Full Product Details