Product Details

SNP ID
rs76253047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35528614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCAAAGCCCTGGACTTGGGGCC[C/T]GCTTCATCTTGGGAAGGGGAGCAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C20orf173 PubMed Links

Gene Details

Gene
C20orf173
Gene Name
chromosome 20 open reading frame 173
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145350.1 641 Intron NP_001138822.1
XM_006723710.2 641 Intron XP_006723773.1
XM_011528577.1 641 UTR 3 XP_011526879.1

View Full Product Details