Product Details
- SNP ID
-
rs16973890
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:37564651 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTTGTGAAGGACATCTAAAGTCTT[C/T]ACCACACTGGACACATGTATAGGGT
- Phenotype
-
MIM: 613903
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF540
PubMed Links
Gene Details
- Gene
- ZNF540
- Gene Name
- zinc finger protein 540
- Gene
- ZNF571
- Gene Name
- zinc finger protein 571
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001290314.1 |
2544 |
Missense Mutation |
AAA,GAA |
K593E |
NP_001277243.1 |
NM_001321272.1 |
2544 |
Missense Mutation |
AAA,GAA |
K593E |
NP_001308201.1 |
NM_016536.4 |
2544 |
Missense Mutation |
AAA,GAA |
K593E |
NP_057620.3 |
XM_011527024.2 |
2544 |
Missense Mutation |
AAA,GAA |
K596E |
XP_011525326.1 |
XM_017026855.1 |
2544 |
Missense Mutation |
AAA,GAA |
K635E |
XP_016882344.1 |
XM_017026856.1 |
2544 |
Missense Mutation |
AAA,GAA |
K596E |
XP_016882345.1 |
XM_017026857.1 |
2544 |
Missense Mutation |
AAA,GAA |
K596E |
XP_016882346.1 |
XM_017026858.1 |
2544 |
Missense Mutation |
AAA,GAA |
K559E |
XP_016882347.1 |
XM_017026859.1 |
2544 |
Missense Mutation |
AAA,GAA |
K559E |
XP_016882348.1 |
XM_017026860.1 |
2544 |
Missense Mutation |
AAA,GAA |
K529E |
XP_016882349.1 |
- Gene
- ZNF571-AS1
- Gene Name
- ZNF571 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details