Product Details

SNP ID
rs77133472
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:183186315 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACCGAGGCGCCGGGCAGCGACCC[C/G]TGCAGCGGAGACAGAGACTGAGCGG
Phenotype
MIM: 150292
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LAMC2 PubMed Links

Gene Details

Gene
LAMC2
Gene Name
laminin subunit gamma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005562.2 40 UTR 5 NP_005553.2
NM_018891.2 40 UTR 5 NP_061486.2
XM_017001273.1 40 UTR 5 XP_016856762.1

View Full Product Details