Product Details

SNP ID
rs10282706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99619801 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAAGCTCTTAGGGAGCTCCAGGA[A/G]CTCTGTCGTCGGTGGCTGAGGCCCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GS1-259H13.2 PubMed Links

Gene Details

Gene
GS1-259H13.2
Gene Name
transmembrane protein 225-like
There are no transcripts associated with this gene.

Gene
ZSCAN25
Gene Name
zinc finger and SCAN domain containing 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145115.2 777 Silent Mutation GAA,GAG E65E NP_660090.2
XM_005250194.2 777 Silent Mutation GAA,GAG E65E XP_005250251.1
XM_011515905.2 777 Silent Mutation GAA,GAG E65E XP_011514207.1
XM_011515906.2 777 Silent Mutation GAA,GAG E65E XP_011514208.1
XM_011515907.2 777 Silent Mutation GAA,GAG E65E XP_011514209.1
XM_011515908.2 777 UTR 5 XP_011514210.1
XM_011515909.2 777 Silent Mutation GAA,GAG E65E XP_011514211.1
XM_011515910.2 777 Silent Mutation GAA,GAG E65E XP_011514212.1
XM_017011824.1 777 Silent Mutation GAA,GAG E65E XP_016867313.1
XM_017011825.1 777 UTR 5 XP_016867314.1
XM_017011826.1 777 UTR 5 XP_016867315.1
XM_017011827.1 777 Silent Mutation GAA,GAG E65E XP_016867316.1

View Full Product Details