Product Details

SNP ID
rs892104687
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99619398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCTGTACCTAATAGTTGCTTAG[C/T]AAATGATTACTATGTTGAATTGACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GS1-259H13.2 PubMed Links

Gene Details

Gene
GS1-259H13.2
Gene Name
transmembrane protein 225-like
There are no transcripts associated with this gene.

Gene
ZSCAN25
Gene Name
zinc finger and SCAN domain containing 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145115.2 Intron NP_660090.2

View Full Product Details