Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_021632.3 | 2059 | UTR 3 | NP_067645.3 | ||
XM_011527196.1 | 2059 | UTR 3 | XP_011525498.1 | ||
XM_017027094.1 | 2059 | UTR 3 | XP_016882583.1 | ||
XM_017027095.1 | 2059 | UTR 3 | XP_016882584.1 | ||
XM_017027096.1 | 2059 | UTR 3 | XP_016882585.1 | ||
XM_017027097.1 | 2059 | UTR 3 | XP_016882586.1 | ||
XM_017027098.1 | 2059 | UTR 3 | XP_016882587.1 | ||
XM_017027099.1 | 2059 | UTR 3 | XP_016882588.1 | ||
XM_017027100.1 | 2059 | UTR 3 | XP_016882589.1 | ||
XM_017027101.1 | 2059 | UTR 3 | XP_016882590.1 | ||
XM_017027102.1 | 2059 | UTR 3 | XP_016882591.1 | ||
XM_017027103.1 | 2059 | UTR 3 | XP_016882592.1 | ||
XM_017027104.1 | 2059 | UTR 3 | XP_016882593.1 |