Product Details

SNP ID
rs113197794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:44873890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGACGGGTCGGTGGCGTTCCCTG[A/T]GCGTTAACGAGTAAACTGTGTCTGT
Phenotype
MIM: 300128
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KDM6A PubMed Links

Gene Details

Gene
KDM6A
Gene Name
lysine demethylase 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291415.1 Intron NP_001278344.1
NM_001291416.1 Intron NP_001278345.1
NM_001291417.1 Intron NP_001278346.1
NM_001291418.1 Intron NP_001278347.1
NM_001291421.1 Intron NP_001278350.1
NM_021140.3 Intron NP_066963.2
XM_005272656.4 Intron XP_005272713.1
XM_005272659.4 Intron XP_005272716.1
XM_011543957.2 Intron XP_011542259.1
XM_011543958.2 Intron XP_011542260.1
XM_011543959.2 Intron XP_011542261.1
XM_011543960.2 Intron XP_011542262.1
XM_011543961.2 Intron XP_011542263.1
XM_011543962.2 Intron XP_011542264.1
XM_011543963.2 Intron XP_011542265.1
XM_011543964.2 Intron XP_011542266.1
XM_011543965.2 Intron XP_011542267.1
XM_011543966.2 Intron XP_011542268.1
XM_011543967.2 Intron XP_011542269.1
XM_011543968.2 Intron XP_011542270.1
XM_011543969.2 Intron XP_011542271.1
XM_011543970.2 Intron XP_011542272.1
XM_011543971.2 Intron XP_011542273.1
XM_011543972.2 Intron XP_011542274.1
XM_011543973.2 Intron XP_011542275.1
XM_011543974.1 Intron XP_011542276.1
XM_011543975.2 Intron XP_011542277.1
XM_017029782.1 Intron XP_016885271.1
XM_017029783.1 Intron XP_016885272.1
XM_017029784.1 Intron XP_016885273.1
XM_017029785.1 Intron XP_016885274.1

View Full Product Details