Product Details

SNP ID
rs10260248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143222638 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTCCACGGAGAAGAAGTACTTCT[A/C]TGAGACCAATATGGTCAACCTGGTA
Phenotype
MIM: 613964
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TAS2R40 PubMed Links

Gene Details

Gene
TAS2R40
Gene Name
taste 2 receptor member 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176882.1 560 Missense Mutation TAT,TCT Y187S NP_795363.1

View Full Product Details