Product Details
- SNP ID
-
hCV25743395
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:48282565 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGCTGGTTTGGAACCGTGTTCTGG[A/G]AAGTGCATTGGAAAATGGGAACTGG
- Phenotype
-
MIM: 603996
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF114
PubMed Links
Gene Details
- Gene
- ZNF114
- Gene Name
- zinc finger protein 114
There are no transcripts associated with this gene.
View Full Product Details