Product Details

SNP ID
hCV25756172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:45110882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCAGTGGTCAGTGTATTTTCACT[A/G]TGGCAACTGTGATGAAAGATTTGGT
Phenotype
MIM: 186947 MIM: 614907
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FKBP3 PubMed Links

Gene Details

Gene
FKBP3
Gene Name
FK506 binding protein 3
There are no transcripts associated with this gene.

Gene
PRPF39
Gene Name
pre-mRNA processing factor 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017922.3 Intron NP_060392.3
Gene
SNORD127
Gene Name
small nucleolar RNA, C/D box 127
There are no transcripts associated with this gene.

View Full Product Details