Product Details

SNP ID
rs35351645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:132583121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCCTTGCCGACTGCAAACCACT[C/T]GCGGCCTGTGCACCCTACTCTAGCA
Phenotype
MIM: 605211 MIM: 616533
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BARHL1 PubMed Links

Gene Details

Gene
BARHL1
Gene Name
BarH like homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020064.3 516 Silent Mutation CTC,CTT L108L NP_064448.1
Gene
CFAP77
Gene Name
cilia and flagella associated protein 77
There are no transcripts associated with this gene.

Gene
DDX31
Gene Name
DEAD-box helicase 31
There are no transcripts associated with this gene.

View Full Product Details