Product Details

SNP ID
rs35037954
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:3022016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCGACCAGCCCCTGCTTGCAAC[A/G]GAGCCCTCCCTGAACCCCAGGGAGA
Phenotype
MIM: 608535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTRT2 PubMed Links

Gene Details

Gene
ACTRT2
Gene Name
actin related protein T2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080431.4 535 Silent Mutation ACA,ACG T110T NP_536356.3

View Full Product Details