Product Details

SNP ID
rs75942973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4523861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTCGTCCACGCAGGCGTGCGC[A/G]TGGGCCACGCGACCCCGGCCCTCGG
Phenotype
MIM: 613247 MIM: 613248
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLIN4 PubMed Links

Gene Details

Gene
PLIN4
Gene Name
perilipin 4
There are no transcripts associated with this gene.

Gene
PLIN5
Gene Name
perilipin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013706.2 1112 Silent Mutation CAC,CAT H353H NP_001013728.2

View Full Product Details