Product Details

SNP ID
rs77162332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123647284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGTTTTGCATTGTAATATGGGT[A/T]TTACTTACCTTGCTTGGAACATGTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KLHL38 PubMed Links

Gene Details

Gene
KLHL38
Gene Name
kelch like family member 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081675.2 Intron NP_001075144.2
XM_005250901.4 Intron XP_005250958.1

View Full Product Details