Product Details

SNP ID
rs75446893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:9288567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTTAAGTTAAGGATTAGAATAT[A/T]ATTAAGTAGAATTTTAAAAGACAAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM41B PubMed Links

Gene Details

Gene
TMEM41B
Gene Name
transmembrane protein 41B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165030.2 Intron NP_001158502.1
NM_015012.3 Intron NP_055827.1

View Full Product Details