Product Details

SNP ID
rs74508691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43475230 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGGCGATCCCGTCGGGGGCAAG[C/G]CACCTGCAACCCAGAACGCAGCAGC
Phenotype
MIM: 609484
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LYPD3 PubMed Links

Gene Details

Gene
LYPD3
Gene Name
LY6/PLAUR domain containing 3
There are no transcripts associated with this gene.

Gene
PHLDB3
Gene Name
pleckstrin homology like domain family B member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198850.3 2534 UTR 3 NP_942147.3
XM_005259169.3 2534 UTR 3 XP_005259226.1
XM_005259170.3 2534 UTR 3 XP_005259227.1
XM_005259171.3 2534 Intron XP_005259228.1
XM_011527236.2 2534 UTR 3 XP_011525538.1

View Full Product Details