Product Details

SNP ID
rs35524230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:77160852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGTCAGTGTAAATGTACTGCTTC[C/T]GACATAAAACGGATGGAAAAAATAA
Phenotype
MIM: 603203
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNG2 PubMed Links

Gene Details

Gene
CCNG2
Gene Name
cyclin G2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004354.2 851 Silent Mutation TCC,TCT S136S NP_004345.1
XM_011532398.1 851 Silent Mutation TCC,TCT S136S XP_011530700.1
XM_011532399.2 851 Silent Mutation TCC,TCT S136S XP_011530701.1

View Full Product Details