Product Details

SNP ID
rs184948109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:36308198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGGCACTGGACCACCTAAATAC[A/G]TTAAAAAAAAATCTGAGATACTATT
Phenotype
MIM: 609431
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MBIP PubMed Links

Gene Details

Gene
MBIP
Gene Name
MAP3K12 binding inhibitory protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144891.1 Intron NP_001138363.1
NM_001308110.1 Intron NP_001295039.1
NM_016586.2 Intron NP_057670.2
XM_005267754.3 Intron XP_005267811.1
XM_005267756.4 Intron XP_005267813.1
XM_017021367.1 Intron XP_016876856.1

View Full Product Details