Product Details

SNP ID
rs62582186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136725921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATCGTTTCCAGAACGAGGCCTGG[A/G]AGTGAGACGCACACTCCTGGTAACC
Phenotype
MIM: 614543
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM69B PubMed Links

Gene Details

Gene
FAM69B
Gene Name
family with sequence similarity 69 member B
There are no transcripts associated with this gene.

Gene
SNHG7
Gene Name
small nucleolar RNA host gene 7
There are no transcripts associated with this gene.

Gene
SNORA17A
Gene Name
small nucleolar RNA, H/ACA box 17A
There are no transcripts associated with this gene.

Gene
SNORA17B
Gene Name
small nucleolar RNA, H/ACA box 17B
There are no transcripts associated with this gene.

View Full Product Details