Product Details
- SNP ID
-
hCV25967742
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:26764433 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAACACGCTCTCGGCAAAGTGAGT[A/C]TGGGCCCTGCCGGGCGTGCGGGCCC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SLC35F6
PubMed Links
Gene Details
- Gene
- SLC35F6
- Gene Name
- solute carrier family 35 member F6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017877.3 |
|
Intron |
|
|
NP_060347.2 |
View Full Product Details