Product Details

SNP ID
rs34156944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132823659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGCTGTTGTTTGGTGCAGGGAG[A/C]CCCTGCCCACATGGCTGGTCAAGCA
Phenotype
MIM: 611179
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC107984115 PubMed Links

Gene Details

Gene
LOC107984115
Gene Name
laforin-like
There are no transcripts associated with this gene.

Gene
SHROOM1
Gene Name
shroom family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172700.1 2000 Silent Mutation NP_001166171.1
NM_133456.2 2000 Silent Mutation NP_597713.2
XM_005271885.4 2000 Silent Mutation XP_005271942.1
XM_005271886.3 2000 Silent Mutation XP_005271943.1
XM_006714534.3 2000 Intron XP_006714597.1
XM_011543167.2 2000 Silent Mutation XP_011541469.1
Gene
SOWAHA
Gene Name
sosondowah ankyrin repeat domain family member A
There are no transcripts associated with this gene.

View Full Product Details