Product Details

SNP ID
rs35672074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140647544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATGCGAATCTCACGCAGGCCCA[C/G]CTTTGCCCCGACTCCTCGACTTGCT
Phenotype
MIM: 600549 MIM: 602137
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IK PubMed Links

Gene Details

Gene
IK
Gene Name
IK cytokine, down-regulator of HLA II
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006083.3 242 Intron NP_006074.2
Gene
MIR3655
Gene Name
microRNA 3655
There are no transcripts associated with this gene.

Gene
NDUFA2
Gene Name
NADH:ubiquinone oxidoreductase subunit A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185012.1 242 Silent Mutation CTG,GTG L14V NP_001171941.1
NM_002488.4 242 Silent Mutation CTG,GTG L14V NP_002479.1
Gene
TMCO6
Gene Name
transmembrane and coiled-coil domains 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300980.1 242 Intron NP_001287909.1
NM_001300982.1 242 Intron NP_001287911.1
NM_018502.4 242 Intron NP_060972.3
XM_005268477.1 242 Intron XP_005268534.1
XM_011537663.2 242 Intron XP_011535965.1
XM_011537665.2 242 Intron XP_011535967.1
XM_011537668.2 242 Intron XP_011535970.1
XM_017009617.1 242 Intron XP_016865106.1
XM_017009618.1 242 Intron XP_016865107.1
XM_017009619.1 242 Intron XP_016865108.1
XM_017009620.1 242 Intron XP_016865109.1

View Full Product Details