Product Details

SNP ID
rs7510795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46050874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGGAAAGGTGGTATCAGGGAG[G/T]CTGAGCATCTGAACCTCCCCAGGGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00899 PubMed Links

Gene Details

Gene
LINC00899
Gene Name
long intergenic non-protein coding RNA 899
There are no transcripts associated with this gene.

Gene
PRR34
Gene Name
proline rich 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018280.2 1421 UTR 3 NP_060750.1
Gene
PRR34-AS1
Gene Name
PRR34 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details