Product Details

SNP ID
rs12999878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88027868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCTTTTTCTTTTTTGTACGCTT[G/A]CGTTCCTCTTTCTTAGAGACTACAT
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
KRCC1 PubMed Links

Gene Details

Gene
KRCC1
Gene Name
lysine rich coiled-coil 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304526.1 1463 Silent Mutation CGC,CGT R232R NP_001291455.1
NM_016618.2 1463 Silent Mutation CGC,CGT R232R NP_057702.1
XM_017004292.1 1463 Silent Mutation CGC,CGT R232R XP_016859781.1

View Full Product Details