Product Details

SNP ID
rs12562288
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:211577020 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTTGGAATAACACCTTGCCGAGT[A/G]GAGTGAGAGGAAAACCAATTACTTA
Phenotype
MIM: 609521
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC30A1 PubMed Links

Gene Details

Gene
SLC30A1
Gene Name
solute carrier family 30 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021194.2 Intron NP_067017.2

View Full Product Details