Product Details

SNP ID
rs16829117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:148648860 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCATGTCATCCTGTGTAGCCTCTT[A/C]TTTTGGACATCCAAACATCTTTTAC
Phenotype
MIM: 611000
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EPC2 PubMed Links

Gene Details

Gene
EPC2
Gene Name
enhancer of polycomb homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015630.3 Intron NP_056445.3
XM_011510941.2 Intron XP_011509243.1
XM_011510943.2 Intron XP_011509245.1

View Full Product Details