Product Details
- SNP ID
-
rs4836471
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:129748391 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGCACAAAAATACGGGGATGGAG[G/T]CTTGTTTCTTTCTCGCTATCTGCCA
- Phenotype
-
MIM: 607513
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ADAMTS19
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116144969] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADAMTS19
- Gene Name
- ADAM metallopeptidase with thrombospondin type 1 motif 19
There are no transcripts associated with this gene.
- Gene
- KIAA1024L
- Gene Name
- KIAA1024 like
View Full Product Details