Product Details

SNP ID
rs11654539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44656117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTCTTTAGCTCTTTCAACCTTC[C/G]ACGACCGGCCTCTCCGCTGCCCTAT
Phenotype
MIM: 616934
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC01180 PubMed Links

Gene Details

Gene
LINC01180
Gene Name
long intergenic non-protein coding RNA 1180
There are no transcripts associated with this gene.

Gene
MEIOC
Gene Name
meiosis specific with coiled-coil domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145080.2 Intron NP_001138552.2
XM_005257236.3 Intron XP_005257293.1

View Full Product Details