Product Details

SNP ID
rs2695444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:156326636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTCTTTTTCTCTACCCCACCCT[A/C]TGGTTTCCCTTCCTCCCTTTCTTTT
Phenotype
MIM: 601828
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NR4A2 PubMed Links

Gene Details

Gene
NR4A2
Gene Name
nuclear receptor subfamily 4 group A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006186.3 Intron NP_006177.1
XM_005246621.3 Intron XP_005246678.1
XM_005246622.3 Intron XP_005246679.1
XM_006712553.3 Intron XP_006712616.1
XM_011511246.1 Intron XP_011509548.1
XM_017004219.1 Intron XP_016859708.1
XM_017004220.1 Intron XP_016859709.1

View Full Product Details