Product Details

SNP ID
rs3183518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70300279 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCAACCATCCCCACTTGCTGGGTA[A/C]TGTCCCAGTTTTAGCATTGAAAGTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LRRC20 PubMed Links

Gene Details

Gene
LRRC20
Gene Name
leucine rich repeat containing 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278211.1 1790 UTR 3 NP_001265140.1
NM_001278212.1 1790 UTR 3 NP_001265141.1
NM_001278213.1 1790 UTR 3 NP_001265142.1
NM_001278214.1 1790 UTR 3 NP_001265143.1
NM_018205.3 1790 UTR 3 NP_060675.1
NM_018239.3 1790 UTR 3 NP_060709.2
NM_207119.2 1790 UTR 3 NP_997002.1
XM_017016374.1 1790 UTR 3 XP_016871863.1
XM_017016375.1 1790 UTR 3 XP_016871864.1

View Full Product Details