Product Details

SNP ID
rs3210817
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:70222677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAGTCTTGTGCTTTTTGGAAGA[A/C]TTATTGGTGGAGAAGTGACAATTCA
Phenotype
MIM: 602010
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SRSF11 PubMed Links

Gene Details

Gene
SRSF11
Gene Name
serine and arginine rich splicing factor 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190987.1 Intron NP_001177916.1
NM_004768.3 Intron NP_004759.1
XM_005271338.3 Intron XP_005271395.1
XM_005271339.3 Intron XP_005271396.1
XM_011542427.2 Intron XP_011540729.1
XM_011542428.2 Intron XP_011540730.1
XM_011542431.2 Intron XP_011540733.1
XM_017002811.1 Intron XP_016858300.1
XM_017002812.1 Intron XP_016858301.1
XM_017002813.1 Intron XP_016858302.1
XM_017002814.1 Intron XP_016858303.1
XM_017002815.1 Intron XP_016858304.1
XM_017002816.1 Intron XP_016858305.1
XM_017002817.1 Intron XP_016858306.1
XM_017002818.1 Intron XP_016858307.1
XM_017002819.1 Intron XP_016858308.1
XM_017002820.1 Intron XP_016858309.1
XM_017002821.1 Intron XP_016858310.1
XM_017002822.1 Intron XP_016858311.1
XM_017002823.1 Intron XP_016858312.1
XM_017002824.1 Intron XP_016858313.1
XM_017002825.1 Intron XP_016858314.1
XM_017002826.1 Intron XP_016858315.1
XM_017002827.1 Intron XP_016858316.1
XM_017002828.1 Intron XP_016858317.1
XM_017002829.1 Intron XP_016858318.1
XM_017002830.1 Intron XP_016858319.1
XM_017002831.1 Intron XP_016858320.1
XM_017002832.1 Intron XP_016858321.1
XM_017002833.1 Intron XP_016858322.1
XM_017002834.1 Intron XP_016858323.1

View Full Product Details