Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004518.5 | 5133 | UTR 3 | NP_004509.2 | ||
NM_172106.2 | 5133 | UTR 3 | NP_742104.1 | ||
NM_172107.3 | 5133 | UTR 3 | NP_742105.1 | ||
NM_172108.4 | 5133 | UTR 3 | NP_742106.1 | ||
NM_172109.2 | 5133 | Intron | NP_742107.1 | ||
XM_011528810.1 | 5133 | UTR 3 | XP_011527112.1 | ||
XM_011528811.1 | 5133 | UTR 3 | XP_011527113.1 | ||
XM_017027841.1 | 5133 | UTR 3 | XP_016883330.1 | ||
XM_017027842.1 | 5133 | UTR 3 | XP_016883331.1 | ||
XM_017027843.1 | 5133 | UTR 3 | XP_016883332.1 | ||
XM_017027844.1 | 5133 | UTR 3 | XP_016883333.1 | ||
XM_017027845.1 | 5133 | UTR 3 | XP_016883334.1 |