Product Details

SNP ID
rs3827085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:23376348 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAACAATCAAGTCCCCTGAGAAAC[A/T]AGTTGGGGTTCTTGCCCACTTGGGC
Phenotype
MIM: 613842 MIM: 611270
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GZF1 PubMed Links

Gene Details

Gene
GZF1
Gene Name
GDNF inducible zinc finger protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317012.1 2054 Intron NP_001303941.1
NM_001317019.1 2054 Intron NP_001303948.1
NM_022482.4 2054 Intron NP_071927.1
Gene
NAPB
Gene Name
NSF attachment protein beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283018.1 2054 UTR 3 NP_001269947.1
NM_001283020.1 2054 UTR 3 NP_001269949.1
NM_001283026.1 2054 UTR 3 NP_001269955.1
NM_022080.2 2054 UTR 3 NP_071363.1
XM_011529313.1 2054 UTR 3 XP_011527615.1
XM_011529315.2 2054 Intron XP_011527617.1
XM_017028008.1 2054 UTR 3 XP_016883497.1

View Full Product Details