Product Details

SNP ID
rs3828332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:179445035 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTGAGTCTTATTCCATGTAAAAC[A/G]TATTTCTAGGTAGCTATTTTCTCCA
Phenotype
MIM: 612344
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF385B PubMed Links

Gene Details

Gene
ZNF385B
Gene Name
zinc finger protein 385B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113397.1 Intron NP_001106868.1
NM_001113398.1 Intron NP_001106869.1
NM_001282725.1 Intron NP_001269654.1
NM_152520.4 Intron NP_689733.3
XM_011510713.2 Intron XP_011509015.1
XM_011510714.2 Intron XP_011509016.1
XM_011510715.2 Intron XP_011509017.1
XM_011510716.2 Intron XP_011509018.1
XM_011510717.2 Intron XP_011509019.1
XM_011510719.2 Intron XP_011509021.1
XM_011510720.2 Intron XP_011509022.1
XM_011510721.2 Intron XP_011509023.1
XM_011510723.2 Intron XP_011509025.1
XM_017003435.1 Intron XP_016858924.1
XM_017003436.1 Intron XP_016858925.1
XM_017003437.1 Intron XP_016858926.1
XM_017003438.1 Intron XP_016858927.1
XM_017003439.1 Intron XP_016858928.1
XM_017003440.1 Intron XP_016858929.1
XM_017003441.1 Intron XP_016858930.1

View Full Product Details